Einzelwafer-Träger
Lieferant: ENTEGRIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H202000011415 H202000021201
ENTEH202000011415EA 0 EUR InStock
ENTEH202000011415 ENTEH202000021201
Einzelwafer-Träger
Wafer-Handling-Zubehör Wafer-Einsätze
Wafer handling trays to safely transport and protect silicon wafers.
  • For internal handling of single wafers
  • Conical pocket holds one wafer facedown contacting only the wafer’s edges
  • Tweezer slots on tray bottom ease wafer removal
  • Stackable for efficient use of storage space, either tray on tray or cover on tray
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge