Graphic mats
Lieferant: ERGOMAT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LB6090 LS6090
ERGOLB6090EA 201 EUR InStock
ERGOLB6090 ERGOLS6090
Graphic mats
Fußmatten Anti-Ermüdungsmatten
These graphic logo mats are extremely strong and durable while being easy to clean and broadly resistant to incidental liquid spills. These mats are promoting a brand and decorate a space with a functional, practical health and safety product that delivers years of relief and aesthetic enjoyment.
  • Free of silicone and latex
  • Moulded in durable, lightweight polyurethane
  • Can be customised with logos or other desired graphics
Available with ergonomic bubble profile or with moderately textured profile and smooth logo: Infinity bubble logo mat is designed to allow workers an anti-fatigue, ergonomic work station. Smooth logo mat offers a flat surface to accommodate pivoting workers and allow trolley accessibility.
Zertifizierungen: Rated to ISO Cleanroom Class 5 (Class 100).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge