Multi-parameter meter, handheld, Eutech CyberScan PC 650
Lieferant: EUTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ECPCWP65043K
EUTEECPCWP65043KEA 1910 EUR InStock
EUTEECPCWP65043K
Multi-parameter meter, handheld, Eutech CyberScan PC 650
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
This portable meter enables users to measure pH, mV, conductivity, TDS, salinity, ions and temperature values. It simultaneously measures and displays up to three parameters.
  • Accepts two-cell and four-cell conductivity probe to measure up to 500 m/s — meter measures pure water
  • Selectable multi-point calibration for more accurate conductivity measurements or single-point calibration for quick, convenient calibration with no mistakes
  • Electrode status report, calibration data and simultaneous display of up to 4 parameters
  • Rugged and IP67-rated waterproof for applications in harsh environments; meter floats on water
  • Recorded readings individually stamped with time and date in GLP compliant format
  • HI-LO limit alarm for easy quality control – Meter alerts when readings fall outside selected ranges
  • Wireless Information Technology: No wires, no cables — Transfer data from meter to computer with the press of a button
Ideal for environmental testing, water quality testing, industrial applications and aquaculture.
Lieferumfang: Supplied with pH electrode ECFC252203B, 4-cell conductivity probe, DAS software, power adapter and carrying kit set (all probes comes with 3 m long cable).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge