Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Fast blue BB salt ≥80% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

M-Clarity™ Qualitätsstufe = MQ200

F3378-1G F3378-25G F3378-5G
F3378-1GEA 39.2 EUR InStock
F3378-1G F3378-25G F3378-5G
Fast blue BB salt ≥80% (Farbstoffgehalt), Sigma-Aldrich®
Fast blue BB salt
Fast Blue BB Salt hemi (zinc chloride) salt, also known as Cl Azoic diazo 20 (Cl 37175) is a diazonium salt that possesses a lipophilic amine lipophilic with an aromatic system of moderate size. It is commercially available as the zinc chloride double salt. It has been used for alkaline phosphatase staining in cells.


Formel: C₃₄H₃₆Cl₄N₆O₆Zn
Melting Pt: 157 °C
Storage Temperature: Gefrierschrank
MDL Number: MFCD00074765
CAS-Nummer: 5486-84-0
EINECS: 226-817-4
Order Now
Spezifikation Testergebnisse
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Turbidity), c = 10 mg/ml; Water Clear to Very Hazy
Solubility (Colour) Yellow to Very Dark Yellow and Brown and Yellow-Brown
Nitrogen ≥8.0%
Dye Content Calculated, Based on Nitrogen Content ≥80%

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge