Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

5-Fluoruracil ≥99% (durch HPLC), Pulver, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ200

F6627-1G F6627-10G F6627-5G
F6627-1GEA 31.9 EUR InStock
F6627-1G F6627-10G F6627-5G
5-Fluoruracil ≥99% (durch HPLC), Pulver, Sigma-Aldrich®
5-Fluoruracil
5-Fluorouracil has been used to induce apoptosis in cells. It has been used as a chemosensitising agent. A potent antitumor agent that affects pyrimidine synthesis by inhibiting thymidylate synthetase, thus depleting intracellular dTTP pools. It is metabolized to ribonucleotides and deoxyribonucleotides, which can be incorporated into RNA and DNA. Treatment of cells with 5-FU leads to an accumulation of cells in S-phase and has been shown to induce p53 dependent apoptosis.


Formel: C₄H₃FN₂O₂
Siedepunkt: 347 °C (1013 hPa)
Melting Pt: 282…286 °C (Zers., Lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00006018
CAS-Nummer: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,04208
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34b30f6a165337be46a329dd264fa290162179a9&vid=34b30f6a165337be46a329dd264fa290162179a9&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour) White to Off-White
Appearance (Form) Powder
Solubility (Colour) Colourless to Very Faint Yellow
Solubility (Turbidity), 50 mg per ml in 1 M Ammonium Hydroxide Clear
Infrared spectrum Conforms to Structure
Purity (HPLC) ≥99%
Recommended Retest Period 2 years

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge