FD0274
FD0275
FD0274EA
63.75
EUR
InStock
FD0274
FD0275
Restriction enzymes, EcoRI, FastDigest™, Fermentas
Nukleinsäure-Reagenzien
Restriktionsenzym
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- Activity (100%) of all FastDigest™ enzymes in one universal reaction buffer
- Direct loading on gels with FastDigest™ Green buffer
- Buffer compatibility (100%) with downstream applications
Designed to complete digestion in 5 to 15 minutes.