Colour Lab Tapes
Lieferant: GA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PAT-13WH PAT-13YE PAT-13LBL PAT-13RE PAT-13GA PAT-13PI PAT-13GR PAT-13OR PAT-13PU PAT-13GS PAT-18WH PAT-18YE PAT-18LBL PAT-18RE PAT-18GA PAT-18PI PAT-18GR PAT-18OR PAT-18PU PAT-18GS PAT-24WH PAT-24YE PAT-24LBL PAT-24RE PAT-24GA PAT-24PI PAT-24GR PAT-24OR PAT-24PU PAT-24GS
GAINPAT-13WHEA 18.2 EUR InStock
GAINPAT-13WH GAINPAT-13YE GAINPAT-13LBL GAINPAT-13RE GAINPAT-13GA GAINPAT-13PI GAINPAT-13GR GAINPAT-13OR GAINPAT-13PU GAINPAT-13GS GAINPAT-18WH GAINPAT-18YE GAINPAT-18LBL GAINPAT-18RE GAINPAT-18GA GAINPAT-18PI GAINPAT-18GR GAINPAT-18OR GAINPAT-18PU GAINPAT-18GS GAINPAT-24WH GAINPAT-24YE GAINPAT-24LBL GAINPAT-24RE GAINPAT-24GA GAINPAT-24PI GAINPAT-24GR GAINPAT-24OR GAINPAT-24PU GAINPAT-24GS
Colour Lab Tapes
Klebebänder
Colour laboratory tape is available in a wide range of colours and widths, for easy identification and colour-coding.
  • Temperature range: –80 to +121 °C
  • Ideal for all general-use applications in the lab
  • Continuous tape format allows use of as much or little as necessary
  • Resistant to gamma irradiation (tested to up to 50 kGy)
  • Available in a wide range of colors and widths, for easy identification and colour-coding
Available in an assortment of eye-catching colours and various widths. Lab tape is perfect for colour-coding and general lab use. Can be written on with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge