Cryogenic warning labels
Lieferant: GA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WL-001-0.25P WL-002-0.25P WL-004-1P WL-008-1R WL-009-1R WL-010-1R WL-011-1P WL-012-1P WL-013-1P WL-015-0.5P WL-016-0.5P WLA-006-0.1P WLA-007-1P
GAINWL-001-0.25PEA 33.5 EUR InStock
GAINWL-001-0.25P GAINWL-002-0.25P GAINWL-004-1P GAINWL-008-1R GAINWL-009-1R GAINWL-010-1R GAINWL-011-1P GAINWL-012-1P GAINWL-013-1P GAINWL-015-0.5P GAINWL-016-0.5P GAINWLA-006-0.1P GAINWLA-007-1P
Cryogenic warning labels
Etiketten
Cryogenic warning labels for storage in liquid nitrogen, freezers, dry ice and other harsh environments. It is resistant to multiple freeze-thaw cycles, boiling water, UV rays, sterilization by gamma irradiation, deep-freeze and harsh lab environments.
  • Water resistant
  • Wear and tear-proof
  • Laminated
  • No perforation
The pictograms in laminated labels are protected against water, alcohol, chemicals and solvents. Temperature range from −196 to +100 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge