The SpinOUT GT-100, GT-600 and GT-1200 columns are versatile, spin-format columns for the desalting and buffer exchange of peptide, protein and nucleic acid solutions ranging from 5l to 4ml sample volumes. Spin-OUT GT-100 is for the purification o 1 * 5 mL
Lieferant: G Bioscienes
The SpinOUT GT-100, GT-600 and GT-1200 columns are versatile, spin-format columns for the desalting and buffer exchange of peptide, protein and nucleic acid solutions ranging from 5l to 4ml sample volumes. Spin-OUT GT-100 is for the purification o 1 * 5 mL
PRODUKT
Erweitert / Kompakt
The SpinOUT™ GT-100, GT-600 and GT-1200 columns are versatile, spin-format columns for the desalting and buffer exchange of peptide, protein and nucleic acid solutions ranging from 5µl to 4ml sample volumes. Spin-OUT™ GT-100 is for the purification o 1 * 5 mL GENO786-868 1 * 5 mL
Hersteller-Artikelnummer:: 786-868
Lokale Artikelnummer:: GENO786-868
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
WEITERE INFORMATIONEN
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder