Taq DNA polymerase, 1000 U
Lieferant: GENSCRIPT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E00008
GENSE00008EA 111 EUR InStock
GENSE00008
Taq DNA polymerase, 1000 U
Nukleinsäure-Reagenzien Enzyme und Kits für Endpunkt-PCR
Taq DNA polymerase is a thermostable DNA polymerase isolated from an E. coli strain that carries the Taq DNA polymerase gene.
  • Used for PCR reactions
  • Terminal transferase activity
  • High purity
One unit is the amount of enzyme that can incorporate 10 nmol of dNTP into acid-insoluble material in 30 minutes at 74 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge