Drucken

Cell Culture Roller Bottles, CELLMASTER®

Lieferant: GREINER BIO ONE
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

680180_18
GREI680180_18EA 216 EUR
GREI680180_18
Cell Culture Roller Bottles, CELLMASTER®
Flaschen Rollerflaschen
PET or PETG with screw caps (HDPE), sterile.

  • Surface suitable for many adherent cell lines
  • PET is very stable and gas permeable; PETG is a glycolised PET which allows more stability at a consistent level of transparency
  • Break-proof
  • All bottles have standard screw caps

Available in short or long form and with a smooth or a ribbed surface. The ribbed design increases the growth area of the roller bottle without changing the height. Short form bottles with a ribbed design have a 2-fold or 2.5-fold expanded growth area. Long forms with a ribbed design have a 5—fold expanded growth area compared to the smooth form. All bottles are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.

Ø×H: Short form (smooth) 116×276 mm; short form (ribbed) 122×275 mm; long form 122×500 mm.

Ø×H: Short form (smooth) 116×276 mm; short form (ribbed) 122×275 mm; long form 122×500 mm

Zertifizierungen: USP Class VI certified

Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Order Now

Learn more