Yeast transformation kit, Fast Yeast Transformation™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
GZ-1
GZ-1EA 557 EUR InStock
GZ-1
Yeast transformation kit, Fast Yeast Transformation™
Nukleinsäure-Reagenzien Transformations-Reagenzien
Fast Yeast Transformation™ is a rapid single step yeast transformation kit that takes less than 10 minutes to prepare competent yeast cells. The competent yeast cells can be used immediately or frozen for later use. This method is suitable for both circular and linear plasmid transformations.
  • High transformation efficiency: 10⁵ to 10⁶ transformants/μg circular DNA
  • Broad spectrum: Compatible with C. albicans, S. pombe, P. pastoris, or S. cerevisiae
  • Transformation procedure takes less than an hour
  • Frozen competent cells are good for use for up to 6 months
  • Simple protocol for multiple plasmid transformation
  • Generates plasmids suitable for multiple downstream applications
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge