H56767.06
H56767.14
H56767.06EA
59.8
EUR
InStock
H56767.06
H56767.14
Sodium phenylpyruvate 98%
Sodium phenylpyruvate
Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.
Purity: 98%
CAS No.: 114-76-1
Molecular Formula: C9H7NaO3
Formula Weight: 186.14
Storage Temperature: 2°C to 8°C
Melting Point: 300°
Beilstein Reference: 4770978
MDL No.: MFCD00002590