H62496.03
H62496.06
H62496.03EA
102
EUR
InStock
H62496.03
H62496.06
Boc-Glu(OMe)-OH 95%
Boc-Glu(OMe)-OH
Purity: 95%
CAS No.: 45214-91-3
Molecular Formula: C11H19NO6
Formula Weight: 261.27
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD00190794