H63895.03
H63895.06
H63895.14
H63895.03EA
125
EUR
InStock
H63895.03
H63895.06
H63895.14
Fmoc-N-Me-D-Leu-OH 97%
Fmoc-N-Me-D-Leu-OH
Purity: 97%
CAS No.: 103478-63-3
Molecular Formula: C22H25NO4
Formula Weight: 367.44
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 7350417
MDL No.: MFCD00235877