Drucken
Lids for 96-well PCR plates, ComfortLid
Lieferant: HAMILTON BONADUZ
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
814300
HAMI814300EA 377 EUR
HAMI814300
Lids for 96-well PCR plates, ComfortLid
Deckel Deckel für Mikrotiterplatten
Gentle pressure is applied to the ComfortLid to seal PCR plates and prevent evaporation and contamination, used in equipped liquid handling workstations.
  • Free of DNase, RNase, pyrogen (endotoxins), human DNA, PCR inhibitors
  • Made of polycarbonate (PC) and thermoplastic elastomers (TPE)
These cost-effective and disposable lid has a stackable design that can be transported by all robotic transport arms.
Verpackung: Packed as 10 lids/blister and 5 blister/box.