HPLC columns, ZORBAX® Extend-C18
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
735700-902 735953-902 746450-902 760450-902 763600-902 763953-902 765600-902 766953-902 773450-902 773700-902 820950-930 821125-930 773450-302 763954-302 5185-5923 735954-302 761753-902 764953-302 727700-902 724700-902 728700-902 728975-902 724975-902 726700-902 726975-902 728975-302 757700-302 757700-902 758700-302 758700-902 766953-302 759700-302 727975-902K 735953-902K 757700-302K 757700-902K 758700-302K 758700-902K 759700-902K 763953-902K 764953-902K 770450-902K 773450-902K 728975-902K
HEWL735700-902EA 637 EUR InStock
HEWL735700-902 HEWL735953-902 HEWL746450-902 HEWL760450-902 HEWL763600-902 HEWL763953-902 HEWL765600-902 HEWL766953-902 HEWL773450-902 HEWL773700-902 HEWL820950-930 HEWL821125-930 HEWL773450-302 HEWL763954-302 HEWL5185-5923 HEWL735954-302 HEWL761753-902 HEWL764953-302 HEWL727700-902 HEWL724700-902 HEWL728700-902 HEWL728975-902 HEWL724975-902 HEWL726700-902 HEWL726975-902 HEWL728975-302 HEWL757700-302 HEWL757700-902 HEWL758700-302 HEWL758700-902 HEWL766953-302 HEWL759700-302 HEWL727975-902K HEWL735953-902K HEWL757700-302K HEWL757700-902K HEWL758700-302K HEWL758700-902K HEWL759700-902K HEWL763953-902K HEWL764953-902K HEWL770450-902K HEWL773450-902K HEWL728975-902K
HPLC columns, ZORBAX® Extend-C18
Chromatographiesäulen
ZORBAX® Extend-C18 columns have high efficiency and a long life at high pH upto pH 11,5 with more effieincy and better peak shape than polymer-based columns.
  • Improve retention, resolution and peak shape of basic compounds
  • High sensitivity for LC/MS separations of peptides and proteins
Extend-C18 columns incorporate a unique patented bidentate silane, combined with a double-endcapping process that protects the silica from dissolution at high pH upto pH 11,5.
Columns are best applied for separations of compounds that are either:
(1) Basic and have little or no retention at low or intermediate pH
(2) More stable or more soluble at high pH
(3) Basic and show poor peak shape at low or intermediate pH
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge