HYDRANAL™ - NEXTGEN Coulomat AG-FI, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
34433-500ML
HONC34433-500MLEA 149.82 EUR InStock
HONC34433-500ML
HYDRANAL™ - NEXTGEN Coulomat AG-FI, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Karl Fischer Titriermittel und Reagenzien
Anolyte for coulometric Karl Fischer titration, for cells with and without diaphragm, Methanol-based, free of Imidazole (free of CMR substances).
  • Imidazole-free
  • Assay of water: 5 - 50 ppm
  • Methanol as solvent
The suffix “FI”, added to the name of the reagent, indicates that the reagent is “free of imidazole”. In 2015, the European Commission classified imidazole as a CMR substance and added H360D statement (“may damage the unborn child”).
Instead of imidazole, an alternative base has been used which does not compromise the performance of the Hydranal reagent.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge