Spike sample standard I for ICP
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E2127-100ML
HONCE2127-100MLEA 702.42 EUR InStock
HONCE2127-100ML
Spike sample standard I for ICP
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
200 mg/L each of Al, As, Ba, Se, Tl, 100 mg/L Fe, 50 mg/L each of Co, Mn, Ni, Pb, Sb, V, Zn, 25 mg/L Cu, 20 mg/L Cr, 5 mg/L each of Ag, Be, Cd in HNO₃ 5%
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge