E2197-100ML
HONCE2197-100MLEA
1078.5
EUR
InStock
HONCE2197-100ML
NexION AFT multi-element solution
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
2 g/L each of Ag, Al, As, Ba, Be, Bi, Ca, Cd, Cr, Co, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn in HNO₃ 2%