Multi-element standard solution 1 for ICP
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E2230-100ML
HONCE2230-100MLEA 144.35 EUR InStock
HONCE2230-100ML
Multi-element standard solution 1 for ICP
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
100 mg/L each of Bi, K, Pb, 50 mg/L each of Al, B, Cr, Li, Mo, Na, Ni, Tl, 10 mg/L each of Ag, Ba, Ca, Cd, Co, Cu, Fe, Mg, Mn, Sr, Zn in HNO₃ 10%
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge