Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E4300-100ML E4300-500ML E4301-100ML E4302-100ML
HONCE4300-100MLEA 248.33 EUR InStock
HONCE4300-100ML HONCE4300-500ML HONCE4301-100ML HONCE4302-100ML
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
Puffer Ionisationspuffer
Zertifizierungen: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)
Lieferumfang: Supplied complete with a detailed Certificate of Analysis (CoA) in every delivery.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge