Circulating baths, for beer force ageing test
Lieferant: HUBER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2041.0001.01
HUBE2041.0001.01EA 14750 EUR InStock
HUBE2041.0001.01
Circulating baths, for beer force ageing test
Umwälzthermostate Geschlossene Umwälzsysteme Temperiersysteme
BFT5 is a special heating and cooling thermostat ideal for beer-forcing test. This unit is fitted with True Adaptive temperature control and programmer for automatic temperature cycles. The beer is artificially ages with changing temperature between 0 and 60 °C in a 24 hour cycle. The unit features a pressure suction pump, programmer with calender and clock function.
  • Working temperature from -40 to +80 °C
  • Pilot ONE® controller with Plug & Play technology,
  • 14,5 cm touch screen and easy menu navigation and graphic capabilities
  • Precise temperature control
  • Over temperature and low liquid level protection
* Cooling capacity at +20; 0 and −20 °C
Zertifizierungen: Units comply with DIN 12876; rated Safety Class III
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge