Solid phase extraction cartridges, HyperSep™ Phenyl
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
60108-386 60108-387 60108-388 60108-389 60108-517 60108-516 60108-707 60108-708 60108-709
HYPE60108-386EA 312 EUR InStock
HYPE60108-386 HYPE60108-387 HYPE60108-388 HYPE60108-389 HYPE60108-517 HYPE60108-516 HYPE60108-707 HYPE60108-708 HYPE60108-709
Solid phase extraction cartridges, HyperSep™ Phenyl
Chromatographiesäulen
Solid phase extraction cartridges, HyperSep™ phenyl offer alternative selectivity for retention of basic compounds. They are used for extraction of aromatic compounds and extraction of basic analytes.
  • Commonly employed for non-polar interactions
  • The electron density of aromatic ring provides different selectivity than other non-polar sorbents
HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Verpackung: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge