Universol™-es liquid scintillation cocktail, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0188248001 0188248004
ICNA0188248001EA 303 EUR InStock
ICNA0188248001 ICNA0188248004
Universol™-es liquid scintillation cocktail, MP Biomedicals
Szintillationsgemische
UniverSol™ is a very efficient environmentally safe, liquid scintillation cocktail of the 'gelling' variety. The physical characteristics of UniverSol™-ES are identical to traditional 'TRITON™ and Toluene' type commercial cocktails.
  • Environmentally safe
  • Non toxic and non flammable
  • Multi-use for aqueous samples
  • High tritium efficiency
  • Both clear and translucent gel phases
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge