Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

5-Brom-2'-deoxyuridin (Broxuridin) ≥99%, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

0210017101 02100171.2 02100171.5 0210017105
ICNA0210017101EA 80.8 EUR InStock
ICNA0210017101 ICNA02100171.2 ICNA02100171.5 ICNA0210017105
5-Brom-2'-deoxyuridin (Broxuridin) ≥99%, weißes Pulver
5-Brom-2'-deoxyuridin (Broxuridin)
5-Bromo-2’-deoxyuridine is a thymidine analog and useful for the study of DNA synthesis where it is incorporated into DNA in place of thymidine. It can be used to label DNA which can be used in conjunction with BrdU Monoclonal Antibody for the immunohistochemical analysis of cell proliferation.

5-Bromo-2'-deoxyuridine (5-BrdU) is a thymidine analogue which is incorporated into DNA. 5-BrdU is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently 5-BrdU is used to study cell signaling and other processes that induce cell proliferation.

Biochem/physiol Actions: Thymidine analog used as a mutagen in genetic research. Selectively incorporated into cellular DNA during S-phase.

Soluble in water (10 mg/ml or 50-100 mg/ml with heat) or sodium hydroxide (50 mg/ml).


Formel: C₉H₁₁BrN₂O₅
Melting Pt: 183…193 °C
Storage Temperature: Gefrierschrank
MDL Number: MFCD00006529
CAS-Nummer: 59-14-3
EINECS: 200-415-9
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
UV/VIS: l max (water) 279 ± 2, 209 ± 3 nm

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge