Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

N-Lauroylsarkosin Natriumsalz ≥94%, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

0219028950 0219028980 0219028990
ICNA0219028950EA 76.2 EUR InStock
ICNA0219028950 ICNA0219028980 ICNA0219028990
N-Lauroylsarkosin Natriumsalz ≥94%, weißes Pulver
N-Lauroylsarkosin Natriumsalz
N-Lauroylsarcosine is an anionic detergent.

N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoproteins; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungistatic agent in aqueous dispersion (1%).

pH: 7.5 - 8.5 (10% aqueous solution)

Sodium Salt: Soluble in water (100 mg/mL - clear to slightly hazy, colorless solution).


Formel: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Storage Temperature: Raumtemperatur
MDL Number: MFCD00042728
CAS-Nummer: 137-16-6
UN: 2811
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0126d80345cce5dd675a83796c379bd44cebacef&vid=0126d80345cce5dd675a83796c379bd44cebacef&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥94%
pH (10% aqueous solution) 7,5 - 8,5
Aggregation Number 2
CMC 14,57 mM at 30 °C (Lit.)
Extinction Coefficient EM = 3 (280 nm) (Lit.)

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge