Laboratory reactor systems, LR-2.ST the High Performer
Lieferant: IKA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
20013291
IKAA20013291EA 19010 EUR InStock
IKAA20013291
Laboratory reactor systems, LR-2.ST the High Performer
Laborreaktor-Systeme
Modular laboratory reactor for the optimisation and reproduction of various chemical reactions, mixing and homogenisation processes on a lab scale. The system is characterised by the agitator mounting, which allows a safe transfer of the higher motor torque.
  • Suitable for vacuum operation
  • Solvent- and temperature-resistant perfluoro elastomer (FFPM) seals come into contact with the sample
  • Infinitely variable speed
  • Torque trend display for measuring changes in viscosity
  • Microprocessor-controlled speed regulation, enables steady speed, also under load
  • Removable WiCo (wireless controller) for remote and safe use in a fume hood
Microprocessor-controlled speed regulation enables steady speed (max. speed 290 min-1). ULTRA-TURRAX® dispersers, temperature sensors, flow breakers and other accessories can be attached to the open ports of the reactor cover.
Bestellinformation: A hose adapter, lock coupling, and reactor vessel are available separately.
Lieferumfang: System includes stand system, EUROSTAR 200 control P4 laboratory stirrer with high torque, safety switch and reactor cover.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge