Butterfly valves, VK series
Lieferant: Welch by Gardner Denver
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
704523 704522
ILMV704523EA 429 EUR InStock
ILMV704523 ILMV704522
Butterfly valves, VK series
Ventile Vakuum-Ventile
These manually operated butterfly valves are made of stainless steel with viton seal.
Leak rate 10-7 mbar L/s
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge