J60741.03
J60741.06
J60741.09
J60741.03EA
68
EUR
InStock
J60741.03
J60741.06
J60741.09
2'-Deoxyguanosine monohydrate
2'-Deoxyguanosine monohydrate
CAS No.: 961-07-9
Molecular Formula: C10H13N5O4
Formula Weight: 267.25
Physical Form: Crystalline powder
Appearance: White to off-white