L05316.22
L05316.36
L05316.22EA
42
EUR
InStock
L05316.22
L05316.36
3-Aminocrotononitril Gemisch aus EZ-Isomeren 96%
3-Aminocrotononitril
Purity: 96%
CAS No.: 1118-61-2
Molecular Formula: C4H6N2
Formula Weight: 82.11
Storage Temperature: 2°C to 8°C
Flash Point: 154°(309°F)
Melting Point: ca 70-75°
Boiling Point: 120°/4mm
Beilstein Reference: 1719815
MDL No.: MFCD00008071