L06526.06
L06526.14
L06526.06EA
43.9
EUR
InStock
L06526.06
L06526.14
DL-Valinmethylesterhydrochlorid 99%
DL-Valine methyl ester hydrochloride
Purity: 99%
CAS No.: 5619-05-6
Molecular Formula: C6H13NO2.HCl
Formula Weight: 167.64
Melting Point: 112-114°
Beilstein Reference: 3912090
MDL No.: MFCD00066118