L09466.14
L09466.06
L09466.03
L09466.14EA
264
EUR
InStock
L09466.14
L09466.06
L09466.03
(2-Bromethyl)cyclohexan ≥99%
(2-Bromethyl)cyclohexan
Purity: 99%
CAS No.: 1647-26-3
Molecular Formula: C8H15Br
Formula Weight: 191.12
Flash Point: 96°(204°F)
Refractive Index: 1.4900
Boiling Point: 70-71°/6mm
Beilstein Reference: 1919816
MDL No.: MFCD00019398