L19244.03
L19244.06
L19244.03EA
59.1
EUR
InStock
L19244.03
L19244.06
3-Brom-4-chlorbenzoesäure ≥97%
3-Bromo-4-chlorobenzoic acid
Purity: 97%
CAS No.: 42860-10-6
Molecular Formula: C7H4BrClO2
Formula Weight: 235.47
Melting Point: 218-222°
Beilstein Reference: 2438796