L19915.06
L19915.06EA
352
EUR
InStock
L19915.06
5-Brom-2-fluor-3-pyridinboronsäure ≥98%
5-Bromo-2-fluoro-3-pyridineboronic acid
Purity: 98%
CAS No.: 501435-91-2
Molecular Formula: C5H4BBrFNO2
Formula Weight: 219.81
Melting Point: 300°
MDL No.: MFCD04039317