Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Leishman's Eosin-Methylenblau für die Histologie, Farbstoff, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

M-Clarity™ Qualitätsstufe = MQ100

L6254-100G L6254-25G
L6254-100GEA 295 EUR InStock
L6254-100G L6254-25G
Leishman's Eosin-Methylenblau für die Histologie, Farbstoff, Sigma-Aldrich®
Leishman's Eosin-Methylenblau
Leishman stain is a Romanowsky-type dye, similar to Wright stain. Leishman stain is used in hematology and tropical medicine to differentiate and identify leukocytes, malarial parasites, and trypanosomes. It is also known as eosin-polychrome methylene blue.

Leishman′s stain has been used for staining blood smears to differentiate blood corpuscles, malarial parasites, trypanosomers, etc. It may be used as an alternative to Giemsa. Leishman’s stain has been used for the identification of macrophage nuclei in cells.It has also been used in hematology study.


Storage Temperature: Room Temperature
MDL Number: MFCD00131498
CAS-Nummer: 12627-53-1
EINECS: 235-732-1
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Dark Green to Very Dark Green
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Colour), c = 1 mg/ml; Methanol Dark Blue to Very Dark Blue
Recommended Retest Period 4 Years

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge