Storage cabinets for gas cylinders, GFL-F90
Lieferant: LACONT UMWELTTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C66-2012-A C66-2014-A C66-2015-A C66-2001-A C66-2002-A C66-2004-A C66-2000-A C66-2010-A
LACOC66-2012-AEA 0 EUR InStock
LACOC66-2012-A LACOC66-2014-A LACOC66-2015-A LACOC66-2001-A LACOC66-2002-A LACOC66-2004-A LACOC66-2000-A LACOC66-2010-A
Storage cabinets for gas cylinders, GFL-F90
Möbel Lagerschränke
Gas cylinder storage system for regular storage outdoors of pressurised gas cylinders.
  • Roof and walls in fire-resistant version
  • Cantilever arm compartment divider with safety chains (fold out)
  • Lockable grilled door
  • Mounted delivery
  • With hinged or double-hinged door depending on version
Zertifizierungen: Conforms with TRGS 510, DIN 4102.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge