LiChrospher® RP-18, HPLC Columns
Lieferant: Hichrom Limited
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LISPRP18-5-50AFEA 784 EUR InStock
LISPRP18-5-50AF LISPRP18-5-10C5 LISPRP18-5125A LISPRP18-5-125AF LISPRP18-5-150AF LISPRP18-5250A LISPRP18-5-250AF LISPRP18-5-250AM LISPRP185300A LISPRP185300AF LISPRP18575AF LISPRP18510C5S
LiChrospher® RP-18, HPLC Columns
Chromatographiesäulen
These LiChrospher® columns are silica available both endcapped and non-endcapped C18 phases. This feature enhances the chromatography selectivity options.
  • Spherical porous silica
  • High efficiency packed columns
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge