VWR® Antibody signal enhancer
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M336-1LEA 0 EUR InStock
M336-1L
VWR® Antibody signal enhancer
Proteinkonjugate und Markierungen
Antibody Signal Enhancer is a Western blotting antibody diluent that increases intensity of protein detection up to 5-fold compared to conventional blotting.
  • Protein-free solution eliminates cross-reaction with antibodies
  • Reduces antigen and reagent use without the need for additional incubation steps
  • Formulated for use with HRP- and biotin-conjugated antibodies and works with both PVDF and nitrocellulose blots
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge