Nitrile gloves, Solo 996
Lieferant: MAPA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
34996559
MAPA34996559EA 34.5 EUR InStock
MAPA34996559
Nitrile gloves, Solo 996
Handschuhe
These disposable nitrile gloves provide excellent mechanical resistance and are ideal for use in oily environments.
  • Excellent dexterity thanks to reduced material thickness
  • Smooth with pebbled fingertips
  • Powdered interior: Easy to put on and to take off
  • Recommended for people who suffer from latex allergies
Typical applications include: Assembly of small oily parts, aeronautics industry, work with composite materials (resins), manufacturing of medicines, pharmaceutical preparation, and research, analysis and handling of precision parts.
Zertifizierungen: EN ISO 374-1 Type B (KPT), EN ISO 374-5; Cat. III
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge