Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Avantor® ACE®, HPLC-/UHPLC-Säulen, Extended Methodenentwicklungs-Kits, 1,7 µm

Lieferant: Advanced Chromatography Technologies

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

MDKE-17-1003UEA 1600 EUR InStock
MDKE-17-1003U MDKE-17-1002U MDKE-17-0503U MDKE-17-0502U MDKE-17-0302U MDKE-17-D1002
Avantor® ACE®, HPLC-/UHPLC-Säulen, Extended Methodenentwicklungs-Kits, 1,7 µm
Chromatographiesäulen
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.

  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 µm and 5 µm ) and complementary phases
  • Analytical IDs in a wide range of column lengths
  • Ideal for method development
  • Compatible with all manufacturers' LC systems

A complete range of kits for all requirements is available, including porous, solid core, HILIC and bioanalytical 300 Å column kits. These kits group together the essential column chemistries for method development and are available in a wide range of column formats.

Maximal empfohlener pH: 2,0 bis 8,0 (1,5 bis 11,5 für SuperC18)

Max. Druck: 1000 bar

Lieferumfang: Avantor® ACE® Extended Methodenentwicklungs-Kits enthalten Avantor® ACE® SuperC18™, Avantor® ACE® C18-Amide und Avantor® ACE® CN-ES Edelstahlsäulen.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge