Divider tote box for shelving systems, Metro®
Lieferant: Metro International
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TB93050BAS TB93030NAT TB92060BAS TB92035BAS TB93060CAS TB93060NAT
METOTB93050BASEA 151 EUR InStock
METOTB93050BAS METOTB93030NAT METOTB92060BAS METOTB92035BAS METOTB93060CAS METOTB93060NAT
Divider tote box for shelving systems, Metro®
Schalen/Schüsseln/Töpfe Einsätze für Schubladen
Divider tote boxes made of polypropylene base resins offer safe and efficient storage of items in bulk, or in compartmentalised fashion with the use of optional dividers and covers. Available in natural gray (NAT), Benstat™ blue static dissipate (BAS) or Bentron™ black conductive (CAS).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge