Hazard BinLiner™ Bags
Lieferant: MICRONOVA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BL4-4060REPC
MICRBL4-4060REPCEA 0 EUR InStock
MICRBL4-4060REPC
Hazard BinLiner™ Bags
Beutel Entsorgungsbeutel
These low density polyethylene Hazard BinLiner™ Bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.
  • Low density polyethylene
  • Hazardous material bags are labeled for waste segregation in the U.S
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Can be used with BLSSH series BinLiner™ Stands
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas
The Biohazard liners are designed for containment and disposal of biomedical waste and the hazardous material liners are designed for segregation and disposal of hazardous waste. Biohazard bags are clearly marked with a Biohazard symbol in black ink.
Zertifizierungen: Certificate of Irradiation and Certificate of Conformance shipped with each order. Validation paperwork available upon request. Minimum irradiation dose: 25 kGy.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge