Surface ATP test swabs, Clean-Trace™
Lieferant: 3M Food Safety
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
UXL100
MMMPUXL100EA 549 EUR InStock
MMMPUXL100
Surface ATP test swabs, Clean-Trace™
Tupfer Tupfer für die Probenahme
Real-time adenosine tri-phosphate (ATP) surface hygiene test which gives an objective indication of cleanliness in seconds.
  • Self contained surface test designed to ensure samples can be collected in hard-to-reach places which are often difficult to clean
  • Metal detectable and blue plastic handle for safe use in a food manufacturing environment
  • A 30-second self-contained sanitation test to assess the standard of food production surfaces
With its unique liquid stable enzyme, the test offers superior levels of repeatability. This aspect of its performance and its incredible simplicity means customers throughout the world have put 3M™ Clean-Trace™ products at the heart of their hygiene monitoring programs. Results measured in the 3M™ Clean-Trace™ Luminometer and provided in Relative Light units (RLU).
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge