Pallets and sumps for drums and containers
Lieferant: asecos
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2516 2520 2521 9843 9844 2500 2504 2508 2510 2512 2514 2501 2505 2509 2511 2513 2515
MSGA2516EA 347 EUR InStock
MSGA2516 MSGA2520 MSGA2521 MSGA9843 MSGA9844 MSGA2500 MSGA2504 MSGA2508 MSGA2510 MSGA2512 MSGA2514 MSGA2501 MSGA2505 MSGA2509 MSGA2511 MSGA2513 MSGA2515
Pallets and sumps for drums and containers
Fasslager
Available either painted (RAL 5010Gentian blue) or hot dip galvanised.
  • Direct storage in the sump pallet or on the grid
  • By adding drummounts, 60-litre and200-litre drums can be stored horizontally
  • Forklift truck pockets
  • Accessible from below on 4 sides
  • Easy and safe in-house transport
Approved for the safe storage of water pollutants and flammable liquids.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge