Available either painted (RAL 5010Gentian blue) or hot dip galvanised.
- Direct storage in the sump pallet or on the grid
- By adding drummounts, 60-litre and200-litre drums can be stored horizontally
- Forklift truck pockets
- Accessible from below on 4 sides
- Easy and safe in-house transport
Approved for the safe storage of water pollutants and flammable liquids.