Methane LB regulator Gas Arc Spec Master regulator (See HPS60GEN datasheet attached) Single-stage, nickel-plated brass, suitable for non-toxic and non-corrosive gases up to N6.0 purity 300 bar max inlet pressure (CGA180 inlet connector to match LB va 1 * 1 ST
Lieferant: MARKET SOURCE PART PROCESS
Methane LB regulator Gas Arc Spec Master regulator (See HPS60GEN datasheet attached) Single-stage, nickel-plated brass, suitable for non-toxic and non-corrosive gases up to N6.0 purity 300 bar max inlet pressure (CGA180 inlet connector to match LB va 1 * 1 ST
PRODUKT
Erweitert / Kompakt
Methane LB regulator Gas Arc Spec Master regulator (See HPS60GEN datasheet attached) Single-stage, nickel-plated brass, suitable for non-toxic and non-corrosive gases up to N6.0 purity 300 bar max inlet pressure (CGA180 inlet connector to match LB va 1 * 1 ST MSPPHPS600-04-03 1 * 1 ST
Hersteller-Artikelnummer:: HPS600-04-03
Lokale Artikelnummer:: MSPPHPS600-04-03
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder