Dropping bottles, APONORM®
Lieferant: MUELLER KREMPEL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
14.015.10 14.015.12 14.015.13 14.015.14 14.015.11
MUEL14.015.10EA 17.7 EUR InStock
MUEL14.015.10 MUEL14.015.12 MUEL14.015.13 MUEL14.015.14 MUEL14.015.11
Dropping bottles, APONORM®
Flaschen Tropfflaschen
Dropper bottles made of amber glass.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Amber coloured to protect light sensitive substances
Thread according to DIN-18 and with dust protection plug, germ-free and hygienic shrink wrap packaging.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge