Identify samples quickly with low profile closures for micro packaging vials. Polypropylene polymer closures are ready to use.
- RNase/DNase free (only sterile closures), non-cytotoxic and non-pyrogenic
Zertifizierungen: Manufactured in compliance with ISO 13485:2003 quality requirements. Micro packaging vials and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Verpackung: Closures are bulk packed without vials, 500 per inner bag, 1000 per case.