Schraubverschlüsse, PPCO, niedriges Profil, 11 mm
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
362821-0112 342821-0110 342821-1111 342821-0112 342821-0114 342821-0115 342821-0116 342821-0118 342821-0111 362821-0111 362821-1111 362821-0114 362821-0116 362821-0110 362821-0115 362821-0118 362821-1112
215-1265EA 162 EUR InStock
215-1265 215-1192 215-1216 215-1210 215-1211 215-1212 215-1213 215-1215 215-1193 215-1264 215-1267 215-1266 215-1653 215-1651 215-1652 215-1654 NALG362821-1112
Schraubverschlüsse, PPCO, niedriges Profil, 11 mm
Verschlüsse Chromatographieampullen-Verschlüsse
Identify samples quickly with low profile closures for micro packaging vials. Polypropylene polymer closures are ready to use.
  • RNase/DNase free (only sterile closures), non-cytotoxic and non-pyrogenic
Zertifizierungen: Manufactured in compliance with ISO 13485:2003 quality requirements. Micro packaging vials and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Verpackung: Closures are bulk packed without vials, 500 per inner bag, 1000 per case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge