Centre well dish for IVF, Nunc™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
150260
NUNC150260EA 318 EUR InStock
NUNC150260
Centre well dish for IVF, Nunc™
Zellkulturschalen Zell- und Gewebekultur-Platten
Non treated surface, sterile (SAL 10⁻⁶), with lid. These IVF centre well dishes meet the exacting requirements of IVF testing, certification, and validation, providing consistently reliable, high quality results during embryo and oocyte washing.
  • Perfect angle of reservoir well walls for locating and easy washing of samples
  • True flat bottom for maintaining optimal temperature
  • Easy grip design for increased safety
  • Certificates of conformity available for every lot
Produced under ISO 13485 with full lot traceability
Zertifizierungen: Produced under ISO 13485 with full lot traceability.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge