Multidishes, poly-D-lysine coated, Nunc™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
152035 152025
735-0227EA 660 EUR InStock
735-0227 NUNC152025
Multidishes, poly-D-lysine coated, Nunc™
Mikrotiterplatten Zellkultur-Platten
PS, with lid. Useful in all areas of cell culture.
  • Raised well rims to lower risk of contamination
  • Excellent optical quality
  • Nunclon™Δ certified surface treatment
Ideal for culturing finicky cells that do not attach to the regular cell culture-treated surfaces.
Bestellinformation: Dishes come packed in sleeves of five.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge